siRNA Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191743.1 | NP_001178672.1 | 13, 14 | 1604 | ||
| XM_008762343.1 | XP_008760565.1 | 14, 15 | 1670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samhd1| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191743.1 | NP_001178672.1 | 13, 14 | 1604 | ||
| XM_008762343.1 | XP_008760565.1 | 14, 15 | 1670 | |||