siRNA Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191743.1 | NP_001178672.1 | 5 | 662 | ||
| XM_008762343.1 | XP_008760565.1 | 6 | 728 | |||
| GenBank mRNA | FN801895.1 | - | 5 | 392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samhd1| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191743.1 | NP_001178672.1 | 5 | 662 | ||
| XM_008762343.1 | XP_008760565.1 | 6 | 728 | |||
| GenBank mRNA | FN801895.1 | - | 5 | 392 | ||