siRNA Details
| Gene Symbol: Fam83d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107796.1 | NP_001101266.1 | 3 | 691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83d| Gene Symbol: Fam83d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107796.1 | NP_001101266.1 | 3 | 691 | ||