siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230849.7 | XP_230849.6 | 6 | 1836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3mbtl1| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230849.7 | XP_230849.6 | 6 | 1836 | ||