siRNA Details
| Gene Symbol: Tox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199392.2 | NP_955424.2 | 5, 6 | 923 | ||
| XM_006235578.2 | XP_006235640.1 | 5, 6 | 957 | |||
| GenBank mRNA | AB096685.1 | - | 5, 6 | 924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tox2| Gene Symbol: Tox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199392.2 | NP_955424.2 | 5, 6 | 923 | ||
| XM_006235578.2 | XP_006235640.1 | 5, 6 | 957 | |||
| GenBank mRNA | AB096685.1 | - | 5, 6 | 924 | ||