siRNA Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044244.2 | NP_001037709.1 | 4 | 528 | ||
| XM_006235585.2 | XP_006235647.1 | 4 | 803 | |||
| GenBank mRNA | BC105768.1 | - | 4 | 529 | 7930355 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm34| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044244.2 | NP_001037709.1 | 4 | 528 | ||
| XM_006235585.2 | XP_006235647.1 | 4 | 803 | |||
| GenBank mRNA | BC105768.1 | - | 4 | 529 | 7930355 | |