siRNA Details
| Gene Symbol: Slc35c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107803.1 | NP_001101273.1 | 2 | 384 | ||
| XM_006235588.2 | XP_006235650.1 | 3 | 1258 | |||
| XM_006235590.2 | XP_006235652.1 | 3 | 1133 | |||
| XM_008762504.1 | XP_008760726.1 | 3 | 1143 | |||
| XM_008762505.1 | XP_008760727.1 | 2 | 2062 | |||
| GenBank mRNA | BC087085.1 | - | 3 | 443 | ||
| BC166837.1 | - | 3 | 427 | 9027110 | ||
| FQ233415.1 | - | 3 | 485 | |||

Slc35c2
Genomic Map