siRNA Details
| Gene Symbol: Slc35c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107803.1 | NP_001101273.1 | 6, 7 | 891 | ||
| XM_006235588.2 | XP_006235650.1 | 7, 8 | 1765 | |||
| XM_006235590.2 | XP_006235652.1 | 7, 8 | 1640 | |||
| XM_008762504.1 | XP_008760726.1 | 7, 8 | 1650 | |||
| XM_008762505.1 | XP_008760727.1 | 6, 7 | 2569 | |||
| GenBank mRNA | BC087085.1 | - | 6, 7 | 1615 | ||
| BC166837.1 | - | 7, 8 | 934 | 9027110 | ||
| FQ233415.1 | - | 7, 8 | 992 | |||

Slc35c2
Genomic Map