siRNA Details
| Gene Symbol: Slc9a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025281.1 | NP_001020452.1 | 11 | 1003 | ||
| XM_006235645.2 | XP_006235707.1 | 10 | 2157 | |||
| XM_006235646.2 | XP_006235708.1 | 11 | 2319 | |||
| GenBank mRNA | AY496958.1 | - | 11 | 1003 | ||
| FQ210940.1 | - | 9 | 759 | |||

Slc9a8
Genomic Map