siRNA Details
| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107805.1 | NP_001101275.1 | 4 | 1556 | ||
| XM_006235653.1 | XP_006235715.1 | 4 | 1550 | |||
| XM_008762507.1 | XP_008760729.1 | 4 | 1550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfatc2| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107805.1 | NP_001101275.1 | 4 | 1556 | ||
| XM_006235653.1 | XP_006235715.1 | 4 | 1550 | |||
| XM_008762507.1 | XP_008760729.1 | 4 | 1550 | |||