siRNA Details
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107806.1 | NP_001101276.1 | 4 | 500 | ||
XM_006235688.2 | XP_006235750.1 | 5 | 725 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107806.1 | NP_001101276.1 | 4 | 500 | ||
XM_006235688.2 | XP_006235750.1 | 5 | 725 |