siRNA Details
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107806.1 | NP_001101276.1 | 7, 8 | 863 | ||
XM_006235688.2 | XP_006235750.1 | 8, 9 | 1088 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107806.1 | NP_001101276.1 | 7, 8 | 863 | ||
XM_006235688.2 | XP_006235750.1 | 8, 9 | 1088 |