siRNA Details
| Gene Symbol: Pmepa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107807.2 | NP_001101277.1 | 3 | 539 | ||
| XM_006235689.1 | XP_006235751.1 | 4 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmepa1| Gene Symbol: Pmepa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107807.2 | NP_001101277.1 | 3 | 539 | ||
| XM_006235689.1 | XP_006235751.1 | 4 | 704 | |||