siRNA Details
| Gene Symbol: Cdh26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191745.1 | NP_001178674.1 | 11 | 1623 | ||
| XM_008762509.1 | XP_008760731.1 | 11 | 1877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh26| Gene Symbol: Cdh26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191745.1 | NP_001178674.1 | 11 | 1623 | ||
| XM_008762509.1 | XP_008760731.1 | 11 | 1877 | |||