siRNA Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173739.1 | NP_001167210.1 | 491 | |||
| GenBank mRNA | BC099155.1 | - | 195 | |||
| CO403993.1 | - | 491 | ||||
| Gene Symbol: LOC103691995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762593.1 | XP_008760815.1 | 5 | 549 | ||

Mrgbp
Genomic Map