siRNA Details
Gene Symbol: Tcfl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762597.1 | XP_008760819.1 | 2 | 639 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
