siRNA Details
| Gene Symbol: Zbtb46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107808.1 | NP_001101278.1 | 2 | 1219 | ||
| XM_008762510.1 | XP_008760732.1 | 2 | 1224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb46| Gene Symbol: Zbtb46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107808.1 | NP_001101278.1 | 2 | 1219 | ||
| XM_008762510.1 | XP_008760732.1 | 2 | 1224 | |||