siRNA Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107811.1 | NP_001101281.1 | 14 | 2374 | ||
| XM_006234354.2 | XP_006234416.1 | 15 | 2520 | |||
| XM_006234355.2 | XP_006234417.1 | 15 | 2516 | |||
| XM_006234356.2 | XP_006234418.1 | 15 | 2520 | |||
| XM_006234357.2 | XP_006234419.1 | 15 | 2520 | |||
| GenBank mRNA | FQ223915.1 | - | 2 | 169 | ||

Gpr155
Genomic Map