siRNA Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107811.1 | NP_001101281.1 | 2 | 343 | ||
| XM_006234354.2 | XP_006234416.1 | 3 | 489 | |||
| XM_006234355.2 | XP_006234417.1 | 3 | 485 | |||
| XM_006234356.2 | XP_006234418.1 | 3 | 489 | |||
| XM_006234357.2 | XP_006234419.1 | 3 | 489 | |||

Gpr155
Genomic Map