siRNA Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025693.1 | NP_001020864.1 | 3 | 454 | ||
| GenBank mRNA | BC098690.1 | - | 3 | 454 | 7386103 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdca7| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025693.1 | NP_001020864.1 | 3 | 454 | ||
| GenBank mRNA | BC098690.1 | - | 3 | 454 | 7386103 | |