siRNA Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761974.1 | XP_008760196.1 | 10 | 3322 | ||
| GenBank mRNA | FQ209892.1 | - | 7 | 494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zak| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761974.1 | XP_008760196.1 | 10 | 3322 | ||
| GenBank mRNA | FQ209892.1 | - | 7 | 494 | ||