siRNA Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107812.2 | NP_001101282.2 | 5 | 586 | ||
| XM_008761908.1 | XP_008760130.1 | 4 | 431 | |||
| GenBank mRNA | BC167092.1 | - | 5 | 513 | ||
| DY575699.1 | - | 4 | 463 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025633, AK005241, AJ414378, BC051534, AK043123, and others.

Metap1d
Genomic Map