siRNA Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107812.2 | NP_001101282.2 | 4 | 526 | ||
| XM_008761908.1 | XP_008760130.1 | 3 | 371 | |||
| GenBank mRNA | BC167092.1 | - | 4 | 453 | ||
| DY575699.1 | - | 3 | 403 | |||

Metap1d
Genomic Map