siRNA Details
| Gene Symbol: Zfp217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107813.1 | NP_001101283.1 | 1 | 1082 | ||
| XM_006235655.2 | XP_006235717.1 | 1 | 1264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp217| Gene Symbol: Zfp217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107813.1 | NP_001101283.1 | 1 | 1082 | ||
| XM_006235655.2 | XP_006235717.1 | 1 | 1264 | |||