siRNA Details
Gene Symbol: Zfp217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107813.1 | NP_001101283.1 | 3 | 1835 | ||
XM_006235655.2 | XP_006235717.1 | 2 | 1900 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107813.1 | NP_001101283.1 | 3 | 1835 | ||
XM_006235655.2 | XP_006235717.1 | 2 | 1900 |