siRNA Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107815.2 | NP_001101285.1 | 7 | 859 | ||
XM_006233594.2 | XP_006233656.1 | 8 | 919 | |||
XM_006233596.2 | XP_006233658.1 | 7 | 763 | |||
XM_008761588.1 | XP_008759810.1 | 8 | 865 | |||
GenBank mRNA | BC169064.1 | - | 7 | 859 | 5599352 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
