siRNA Details
| Gene Symbol: Traf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107815.2 | NP_001101285.1 | 7 | 973 | ||
| XM_006233594.2 | XP_006233656.1 | 8 | 1033 | |||
| XM_006233596.2 | XP_006233658.1 | 7 | 877 | |||
| XM_008761588.1 | XP_008759810.1 | 8 | 979 | |||
| GenBank mRNA | BC169064.1 | - | 7 | 973 | 5599352 | |

Traf2
Genomic Map