siRNA Details
| Gene Symbol: Zmynd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198770.2 | NP_942065.1 | 5, 6 | 733 | ||
| XM_006233597.2 | XP_006233659.1 | 731 | ||||
| GenBank mRNA | AJ488144.1 | - | 564 | |||
| BC088093.1 | - | 5, 6 | 733 | 7368043 | ||
| Gene Symbol: LOC100910854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253280.2 | XP_006253342.1 | 1, 2 | 694 | ||

Zmynd19
Genomic Map