siRNA Details
| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025694.2 | NP_001020865.1 | 7 | 811 | ||
| XM_008761589.1 | XP_008759811.1 | 8 | 794 | |||
| XM_008761590.1 | XP_008759812.1 | 8 | 786 | |||
| XM_008761591.1 | XP_008759813.1 | 7 | 734 | |||
| XM_008761592.1 | XP_008759814.1 | 7 | 610 | |||
| XM_008761593.1 | XP_008759815.1 | 6 | 649 | |||
| XM_008761594.1 | XP_008759816.1 | 5 | 467 | |||
| XM_008761595.1 | XP_008759817.1 | 4 | 413 | |||
| XM_008761596.1 | XP_008759818.1 | 8 | 864 | |||
| GenBank mRNA | BC097396.1 | - | 7 | 812 | 7453380 | |

Fam166a
Genomic Map