siRNA Details
| Gene Symbol: Ssna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233613.1 | XP_006233675.1 | 3 | 508 | ||
| GenBank mRNA | BC166994.1 | - | 3 | 424 | 7304458 | |
| CB576781.1 | - | 3 | 461 | |||
| FN805842.1 | - | 3 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssna1| Gene Symbol: Ssna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233613.1 | XP_006233675.1 | 3 | 508 | ||
| GenBank mRNA | BC166994.1 | - | 3 | 424 | 7304458 | |
| CB576781.1 | - | 3 | 461 | |||
| FN805842.1 | - | 3 | 403 | |||