siRNA Details
| Gene Symbol: Rexo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033884.1 | NP_001029056.1 | 6 | 1101 | ||
| GenBank mRNA | BC161820.1 | - | 6 | 1066 | 9088929 | |
| CB800860.1 | - | 1 | 56 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rexo4| Gene Symbol: Rexo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033884.1 | NP_001029056.1 | 6 | 1101 | ||
| GenBank mRNA | BC161820.1 | - | 6 | 1066 | 9088929 | |
| CB800860.1 | - | 1 | 56 | |||