siRNA Details
| Gene Symbol: Ddx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107824.2 | NP_001101294.2 | 10 | 928 | ||
| XM_006233799.2 | XP_006233861.1 | 9 | 1100 | |||
| GenBank mRNA | BC160917.1 | - | 10 | 928 | 7457829 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx31| Gene Symbol: Ddx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107824.2 | NP_001101294.2 | 10 | 928 | ||
| XM_006233799.2 | XP_006233861.1 | 9 | 1100 | |||
| GenBank mRNA | BC160917.1 | - | 10 | 928 | 7457829 | |