siRNA Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107825.1 | NP_001101295.1 | 2 | 460 | ||
| XM_006233800.2 | XP_006233862.1 | 2 | 460 | |||
| XM_006233802.2 | XP_006233864.1 | 2 | 769 | |||
| XM_006233804.2 | XP_006233866.1 | 2 | 460 | |||

Ntng2
Genomic Map