siRNA Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100706.1 | NP_001094176.1 | 7 | 1299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc27a4| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100706.1 | NP_001094176.1 | 7 | 1299 | ||