siRNA Details
| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137562.2 | NP_001131034.2 | 3 | 160 | ||
| GenBank mRNA | BC169094.1 | - | 3 | 150 | 7933748 | |
| CB313752.1 | - | 3 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urm1| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137562.2 | NP_001131034.2 | 3 | 160 | ||
| GenBank mRNA | BC169094.1 | - | 3 | 150 | 7933748 | |
| CB313752.1 | - | 3 | 163 | |||