siRNA Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009692.1 | NP_001009692.1 | 5 | 623 | ||
| XM_006233816.2 | XP_006233878.1 | 5 | 600 | |||
| XM_006233817.2 | XP_006233879.1 | 5 | 600 | |||
| XM_006233818.2 | XP_006233880.1 | 5 | 599 | |||
| XM_006233819.2 | XP_006233881.1 | 5 | 599 | |||
| XM_006233820.2 | XP_006233882.1 | 5 | 599 | |||
| XM_006233822.2 | XP_006233884.1 | 5 | 599 | |||
| XM_008761661.1 | XP_008759883.1 | 5 | 600 | |||
| XM_008761662.1 | XP_008759884.1 | 5 | 599 | |||
| GenBank mRNA | BC087652.1 | - | 5 | 623 | 7312960 | |

Sh3glb2
Genomic Map