siRNA Details
| Gene Symbol: Crat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004085.2 | NP_001004085.1 | 9 | 1375 | ||
| XM_006233823.1 | XP_006233885.1 | 10 | 1895 | |||
| XM_008761663.1 | XP_008759885.1 | 10 | 1378 | |||
| XM_008761664.1 | XP_008759886.1 | 9 | 1244 | |||
| GenBank mRNA | AJ620886.1 | - | 9 | 1153 | ||
| BC083616.1 | - | 9 | 1375 | 7133847 | ||
| FQ230824.1 | - | 10 | 1490 | |||

Crat
Genomic Map