siRNA Details
| Gene Symbol: RGD1311084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100979.1 | NP_001094449.1 | 3 | 937 | ||
| XM_006233913.2 | XP_006233975.1 | 4 | 911 | |||
| XM_006233914.2 | XP_006233976.1 | 4 | 911 | |||
| GenBank mRNA | BC127535.1 | - | 3 | 937 | 7133909 | |
| FQ214464.1 | - | 3 | 682 | |||

RGD1311084
Genomic Map