siRNA Details
| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039197.1 | NP_001034286.1 | 2 | 389 | ||
| GenBank mRNA | BC111704.1 | - | 2 | 389 | 7936904 | |
| FQ222570.1 | - | 2 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor1b| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039197.1 | NP_001034286.1 | 2 | 389 | ||
| GenBank mRNA | BC111704.1 | - | 2 | 389 | 7936904 | |
| FQ222570.1 | - | 2 | 449 | |||