siRNA Details
| Gene Symbol: LOC102554659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233926.2 | XP_006233988.1 | 5 | 1267 | ||
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107830.1 | NP_001101300.1 | 5 | 1184 | ||
| XM_008761668.1 | XP_008759890.1 | 5 | 1267 | |||
| XM_008761669.1 | XP_008759891.1 | 5 | 1267 | |||

LOC102554659
Genomic Map