siRNA Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107830.1 | NP_001101300.1 | 17 | 3066 | ||
| XM_008761668.1 | XP_008759890.1 | 17 | 3149 | |||
| XM_008761669.1 | XP_008759891.1 | 17 | 3149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamc3| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107830.1 | NP_001101300.1 | 17 | 3066 | ||
| XM_008761668.1 | XP_008759890.1 | 17 | 3149 | |||
| XM_008761669.1 | XP_008759891.1 | 17 | 3149 | |||