siRNA Details
| Gene Symbol: Ptges2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107832.1 | NP_001101302.1 | 2 | 923 | ||
| GenBank mRNA | FQ231959.1 | - | 2 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptges2| Gene Symbol: Ptges2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107832.1 | NP_001101302.1 | 2 | 923 | ||
| GenBank mRNA | FQ231959.1 | - | 2 | 416 | ||