siRNA Details
| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107834.1 | NP_001101304.1 | 5 | 877 | ||
| XM_006233975.2 | XP_006234037.1 | 5 | 939 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbx3| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107834.1 | NP_001101304.1 | 5 | 877 | ||
| XM_006233975.2 | XP_006234037.1 | 5 | 939 | |||