siRNA Details
| Gene Symbol: Fbxw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107835.2 | NP_001101305.1 | 2 | 408 | ||
| XM_006233976.1 | XP_006234038.1 | 2 | 542 | |||
| XM_006233977.2 | XP_006234039.1 | 3 | 458 | |||
| XM_006233980.2 | XP_006234042.1 | 3 | 366 | |||
| XM_008761750.1 | XP_008759972.1 | 3 | 2017 | |||
| XM_008761751.1 | XP_008759973.1 | 4 | 623 | |||
| XM_008761752.1 | XP_008759974.1 | 2 | 344 | |||
| GenBank mRNA | BC166990.1 | - | 2 | 408 | 7110795 | |

Fbxw2
Genomic Map