siRNA Details
| Gene Symbol: Cntrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234052.2 | XP_006234114.1 | 34, 35 | 5788 | ||
| XM_006234055.2 | XP_006234117.1 | 34, 35 | 5788 | |||
| XM_008761801.1 | XP_008760023.1 | 34, 35 | 5752 | |||
| XM_008761802.1 | XP_008760024.1 | 33, 34 | 5608 | |||
| XM_008761803.1 | XP_008760025.1 | 24, 25 | 4363 | |||
| XM_008761804.1 | XP_008760026.1 | 26, 27 | 4259 | |||

Cntrl
Genomic Map