siRNA Details
| Gene Symbol: Cntrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234052.2 | XP_006234114.1 | 4 | 606 | ||
| XM_006234055.2 | XP_006234117.1 | 4 | 606 | |||
| XM_008761801.1 | XP_008760023.1 | 4 | 606 | |||
| XM_008761802.1 | XP_008760024.1 | 4 | 606 | |||

Cntrl
Genomic Map