siRNA Details
| Gene Symbol: Rc3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107839.1 | NP_001101309.1 | 17, 18 | 3078 | ||
| XM_008761754.1 | XP_008759976.1 | 17, 18 | 3179 | |||
| XM_008761755.1 | XP_008759977.1 | 16, 17 | 2873 | |||

Rc3h2
Genomic Map