siRNA Details
Gene Symbol: Zbtb26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107840.1 | NP_001101310.1 | 2 | 1253 | ||
XM_006234082.2 | XP_006234144.1 | 1 | 1125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107840.1 | NP_001101310.1 | 2 | 1253 | ||
XM_006234082.2 | XP_006234144.1 | 1 | 1125 |