siRNA Details
| Gene Symbol: Rsbn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135872.1 | NP_001129344.1 | 7 | 2276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsbn1l| Gene Symbol: Rsbn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135872.1 | NP_001129344.1 | 7 | 2276 | ||