siRNA Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191748.1 | NP_001178677.1 | 1133 | |||
| Gene Symbol: LOC103690019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762747.1 | XP_008760969.1 | 11 | 1136 | ||

Dbf4
Genomic Map